This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01528387_g1
See other GPX5 GE Assays ›
Gene Symbol:
Gpx5
Gene Name:
glutathione peroxidase 5
Gene Aliases:
-
Chromosome Location:
Chr.17: 45508657 - 45518686 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01528387_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105738.1 NP_001099208.1 386 107
GenBank mRNA X62404.1 - 431 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700000695.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
cellular response to oxidative stress
oxidation-reduction process
glutathione peroxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700004756.rn.1 TC1700000695.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0