This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01528424_m1
See other GPX6 GE Assays ›
Gene Symbol:
Gpx6
Gene Name:
glutathione peroxidase 6
Gene Aliases:
OBPII, Ry2d1
Chromosome Location:
Chr.17: 45439268 - 45446952 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01528424_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_147165.1 NP_671694.1 4 - 5 508 122
GenBank mRNA M76733.1 - 4 - 5 508 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700001845.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpx6

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
oxidation-reduction process
glutathione peroxidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119184 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 503988 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 675711 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 579188 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 167982 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119184 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119184 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119184 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P125974 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P571388 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 61038 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10798534 - -
Affymetrix Rat Gene 1.0 ST Array 10798535 - -
Affymetrix Rat Gene 1.0 ST Array 10798531 - -
Affymetrix Rat Gene 1.0 ST Array 10798530 - -
Affymetrix Rat Gene 1.0 ST Array 10798532 - -
Affymetrix Rat Gene 1.0 ST Array 10798533 - -
Affymetrix Rat Gene 1.1 ST Array 10798529 - -
Rat Genome 230 2.0 Array 1388085_PM_at - -
Rat Genome 230 2.0 Array 1388085_at - -
Affymetrix HT Rat Focus Array 1388085_at - -
Affymetrix HT Rat Focus Array 1388085_PM_at - -
Affymetrix Rat Clariom S Assay PSR1700012566.rn.1 TC1700001845.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700012568.rn.1 TC1700001845.rn.1 RTA TC
Rat Expression Set 230 1388085_PM_at - -
Rat Expression Set 230 1388085_at - -
Affymetrix Rat Gene 1.0 ST Array 10798529 - -
Rat Genome U34 Set M76733_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22172436 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359971 - -
RatRef-12 Expression BeadChip ILMN_56172 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline