This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01528713_g1
See other MSANTD4 GE Assays ›
Gene Symbol:
Msantd4
Gene Name:
Myb/SANT-like DNA-binding domain containing 4 with coiled-coils
Gene Aliases:
-
Chromosome Location:
Chr.8: 1503018 - 1513626 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01528713_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msantd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024357.1 NP_001019528.1 2 - 3 656 71
GenBank mRNA BC096000.1 - 2 - 3 656 7308051 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000007.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Msantd4

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P114183 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 184321 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 118769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 230228 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 165258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611647 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 972462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 341308 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 622860 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 300130 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 167585 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114183 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P114183 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P114183 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10907813 - -
Affymetrix Rat Gene 1.0 ST Array 10907814 - -
Affymetrix Rat Gene 1.0 ST Array 10907812 - -
Affymetrix Rat Gene 1.1 ST Array 10907811 - -
Rat Genome 230 2.0 Array 1375884_PM_at - -
Rat Genome 230 2.0 Array 1375884_at - -
Affymetrix HT Rat Focus Array 1375884_PM_at - -
Affymetrix HT Rat Focus Array 1375884_at - -
Affymetrix Rat Clariom S Assay PSR0800000037.rn.1 TC0800000007.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800000040.rn.1 TC0800000007.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800000038.rn.1 TC0800000007.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800000039.rn.1 TC0800000007.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800000035.rn.1 TC0800000007.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800000036.rn.1 TC0800000007.rn.1 RTA TC
Rat Expression Set 230 1375884_at - -
Rat Expression Set 230 1375884_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10907811 - -
Rat Genome U34 Set rc_AI059305_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21932906 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1373488 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0