This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01529538_g1
See other NABP2 GE Assays ›
Gene Symbol:
Nabp2
Gene Name:
nucleic acid binding protein 2
Gene Aliases:
Obfc2b, RGD1308158
Chromosome Location:
Chr.7: 2820006 - 2825619 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01529538_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nabp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034939.1 NP_001030111.2 4 - 5 771 94
NM_001244819.1 NP_001231748.1 5 - 6 494 94
XM_006240769.2 XP_006240831.1 5 - 6 735 94
XM_006240770.2 XP_006240832.1 5 - 6 590 94
XM_006240771.2 XP_006240833.1 5 - 6 585 94
XM_008765029.1 XP_008763251.1 5 - 6 750 94
GenBank mRNA BC104710.1 - 4 - 5 771 7319804 94
DV214819.1 - 5 - 6 494 94
FQ219805.1 - 5 - 6 482 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700001992.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nabp2

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
mitotic cell cycle checkpoint
response to ionizing radiation
single-stranded DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700015521.rn.1 TC0700001992.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0