This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01529810_g1
See other NAA10 GE Assays ›
Gene Symbol:
Naa10
Gene Name:
N(alpha)-acetyltransferase 10, NatA catalytic subunit
Gene Aliases:
Ard1, Ard1a, RGD1565315
Chromosome Location:
Chr.X: 156863645 - 156868950 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01529810_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135839.1 NP_001129311.1 2 - 3 263 89
XM_006229591.2 XP_006229653.1 2 - 3 573 89
XM_006229592.2 XP_006229654.1 2 - 3 372 89
GenBank mRNA FQ235221.1 - 2 - 3 207 89

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
internal protein amino acid acetylation
N-terminal peptidyl-serine acetylation
N-terminal peptidyl-glutamic acid acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
acetyltransferase activity
ribosome binding
peptide-serine-N-acetyltransferase activity
peptide-glutamate-N-acetyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0X00007942.rn.1 TC0X00001306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00007943.rn.1 TC0X00001306.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0X00007941.rn.1 TC0X00001306.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0