This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01531265_g1
See other MTF2 GE Assays ›
Gene Symbol:
Mtf2
Gene Name:
metal response element binding transcription factor 2
Gene Aliases:
RGD1304727
Chromosome Location:
Chr.14: 2656777 - 2701024 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01531265_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100898.1 NP_001094368.1 4 - 5 612 151
XM_006250569.2 XP_006250631.1 5 - 6 807 151
XM_008769939.1 XP_008768161.1 4 - 5 517 151
GenBank mRNA BC093403.1 - 4 - 5 572 151
BC160928.1 - 2 - 3 181 151
FQ227583.1 - 5 - 6 809 151

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400001249.rn.1

Additional Information:

This assay is in the 5'UTR region of the XM_006250569.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
segment specification
biological_process
stem cell population maintenance
positive regulation of transcription from RNA polymerase II promoter
stem cell differentiation
negative regulation of histone H3-K27 methylation
positive regulation of histone H3-K27 methylation
molecular_function
zinc ion binding
methylated histone binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400009296.rn.1 TC1400001249.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400009297.rn.1 TC1400001249.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400009294.rn.1 TC1400001249.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21908791 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0