This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01531410_m1
See other TMLHE GE Assays ›
Gene Symbol:
Tmlhe
Gene Name:
trimethyllysine hydroxylase, epsilon
Gene Aliases:
Tmlh
Chromosome Location:
Chr.20: 237461 - 285125 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01531410_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmlhe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133387.2 NP_596878.2 4 - 5 480 75
XM_006255855.1 XP_006255917.1 3 - 4 489 75
GenBank mRNA AF374406.1 - 2 - 3 321 75
BC085923.1 - 4 - 5 480 7109209 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000000005.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmlhe

Panther Classification:

Molecular Function -

hydroxylase

Gene Ontology Categories:

Function(s) Process(es)

carnitine biosynthetic process
negative regulation of oxidoreductase activity
oxidation-reduction process
iron ion binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
L-ascorbic acid binding
trimethyllysine dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000000060.rn.1 TC2000000005.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000000065.rn.1 TC2000000005.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000000056.rn.1 TC2000000005.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000000061.rn.1 TC2000000005.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR2000000067.rn.1 TC2000000005.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline