This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01532005_m1
See other ATAT1 GE Assays ›
Gene Symbol:
Atat1
Gene Name:
alpha tubulin acetyltransferase 1
Gene Aliases:
3110080J08Rik, Mec17, RGD1303066, TAT
Chromosome Location:
Chr.20: 3351268 - 3364565 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01532005_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212498.1 NP_997663.1 6 - 7 550 64
XM_006255948.2 XP_006256010.1 6 - 7 585 64
XM_006255950.2 XP_006256012.1 6 - 7 585 64
XM_006255951.2 XP_006256013.1 6 - 7 585 64
XM_008772731.1 XP_008770953.1 6 - 7 585 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000000102.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atat1

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
dentate gyrus development
regulation of fat cell differentiation
neuron development
regulation of microtubule cytoskeleton organization
alpha-tubulin acetylation
positive regulation of NLRP3 inflammasome complex assembly
tubulin N-acetyltransferase activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000000670.rn.1 TC2000000102.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0