This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01532633_m1
See other PPP1R1A GE Assays ›
Gene Symbol:
Ppp1r1a
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 1A
Gene Aliases:
-
Chromosome Location:
Chr.7: 145146480 - 145154146 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01532633_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022676.3 NP_073167.1 1 - 2 218 113
XM_006242437.2 XP_006242499.1 1 - 2 233 113
GenBank mRNA AJ276593.1 - 1 - 2 91 113
AY648296.1 - 1 - 2 218 113
BC078820.1 - 1 - 2 219 7127505 113
FQ217772.1 - 1 - 2 187 113
J05592.1 - 1 - 2 90 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700004006.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r1a

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
negative regulation of protein kinase activity
intracellular signal transduction
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700032464.rn.1 TC0700004006.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700032460.rn.1 TC0700004006.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700032463.rn.1 TC0700004006.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0