This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01534699_m1
See other TNFRSF19 GE Assays ›
Gene Symbol:
Tnfrsf19
Gene Name:
tumor necrosis factor receptor superfamily, member 19
Gene Aliases:
RGD1564996
Chromosome Location:
Chr.15: 41249820 - 41316203 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01534699_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044229.2 NP_001037694.1 7 - 8 838 60
GenBank mRNA CO396223.1 - 3 - 4 384 60
DQ466087.1 - 7 - 8 838 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500001828.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Gene Ontology Categories:

Function(s)

hair follicle development
positive regulation of I-kappaB kinase/NF-kappaB signaling

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000141 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152602 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 28384 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 155722 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 266476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 720687 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 402344 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 706700 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 344977 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 963052 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587670 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 698540 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 612443 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 434357 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 484809 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 846058 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 165153 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 777480 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657437 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 166201 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 492583 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 208084 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 814129 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325337 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 279218 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 659095 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 327229 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 101219 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 306496 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 139486 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 265490 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 855528 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773020 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 71462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 712640 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 599617 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 30410 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 392369 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894913 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 709335 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 952185 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 511812 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 286883 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 407565 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 275533 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 782923 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 231471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 588478 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 626011 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 64197 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112219 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 76720 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 44566 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 134898 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168231 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 106155 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 123086 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000141 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000141 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000141 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100561 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 41038 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 70327 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 4952 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 66701 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 54944 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10784343 - -
Affymetrix Rat Gene 1.0 ST Array 10784336 - -
Affymetrix Rat Gene 1.0 ST Array 10784341 - -
Affymetrix Rat Gene 1.0 ST Array 10784338 - -
Affymetrix Rat Gene 1.0 ST Array 10784337 - -
Affymetrix Rat Gene 1.0 ST Array 10784339 - -
Affymetrix Rat Gene 1.0 ST Array 10784342 - -
Affymetrix Rat Gene 1.0 ST Array 10784335 - -
Affymetrix Rat Gene 1.0 ST Array 10784340 - -
Affymetrix Rat Gene 1.1 ST Array 10784334 - -
Rat Genome 230 2.0 Array 1393591_PM_at - -
Rat Genome 230 2.0 Array 1393591_at - -
Affymetrix HT Rat Focus Array 1393591_at - -
Affymetrix HT Rat Focus Array 1393591_PM_at - -
Affymetrix Rat Clariom S Assay PSR1500012274.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012276.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012271.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012275.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012267.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012269.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012270.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012273.rn.1 TC1500001828.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500012272.rn.1 TC1500001828.rn.1 RTA TC
Rat Expression Set 230 1393591_PM_at - -
Rat Expression Set 230 1393591_at - -
Affymetrix Rat Gene 1.0 ST Array 10784334 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1369872 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0