This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01536332_m1
See other DUSP15 GE Assays ›
Gene Symbol:
Dusp15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
-
Chromosome Location:
Chr.3: 148417990 - 148428494 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01536332_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108598.2 NP_001102068.2 3 - 4 345 93
NM_001244784.1 NP_001231713.1 3 - 4 345 93
GenBank mRNA BC168211.1 - 3 - 4 174 7135378 93
CB576376.1 - 3 - 4 222 93
CB585518.1 - 3 - 4 345 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300004497.rn.1

Set Membership:

Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
transforming growth factor beta receptor signaling pathway
peptidyl-tyrosine dephosphorylation
regulation of cell proliferation
positive regulation of JNK cascade
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300034442.rn.1 TC0300004497.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0