This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01536859_g1
See other POLR2C GE Assays ›
Gene Symbol:
Polr2c
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide C
Gene Aliases:
-
Chromosome Location:
Chr.19: 10575060 - 10581842 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01536859_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012473.1 NP_001012491.1 7 - 8 636 75
GenBank mRNA BC089905.1 - 7 - 8 636 7322752 75
FQ218345.1 - 7 - 8 553 75
FQ225242.1 - 7 - 8 656 75
FQ226282.1 - 7 - 8 677 75
FQ235323.1 - 7 - 8 675 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000987.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2c

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
RNA polymerase II activity
DNA binding
protein dimerization activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321329 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 45487 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 30517 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 686631 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 395422 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 12296 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321329 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321329 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321329 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321329 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10809244 - -
Affymetrix Rat Gene 1.0 ST Array 10809243 - -
Affymetrix Rat Gene 1.0 ST Array 10809246 - -
Affymetrix Rat Gene 1.0 ST Array 10809242 - -
Affymetrix Rat Gene 1.0 ST Array 10809249 - -
Affymetrix Rat Gene 1.0 ST Array 10809247 - -
Affymetrix Rat Gene 1.0 ST Array 10809245 - -
Affymetrix Rat Gene 1.0 ST Array 10809248 - -
Affymetrix Rat Gene 1.0 ST Array 10809250 - -
Affymetrix Rat Gene 1.1 ST Array 10809240 - -
Rat Genome 230 2.0 Array 1390491_PM_at - -
Rat Genome 230 2.0 Array 1398899_at - -
Rat Genome 230 2.0 Array 1388570_at - -
Rat Genome 230 2.0 Array 1372300_PM_at - -
Rat Genome 230 2.0 Array 1390491_at - -
Rat Genome 230 2.0 Array 1372300_at - -
Rat Genome 230 2.0 Array 1388570_PM_at - -
Rat Genome 230 2.0 Array 1398899_PM_at - -
Affymetrix HT Rat Focus Array 1398899_at - -
Affymetrix HT Rat Focus Array 1398899_PM_at - -
Affymetrix Rat Clariom S Assay PSR1900007869.rn.1 TC1900000987.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007866.rn.1 TC1900000987.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900007868.rn.1 TC1900000987.rn.1 RTA TC
Rat Expression Set 230 1398899_PM_at - -
Rat Expression Set 230 1398899_at - -
Affymetrix Rat Gene 1.0 ST Array 10809240 - -
Rat Genome U34 Set rc_AI170414_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22009757 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0