This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01537196_m1
See other DUSP13 GE Assays ›
Gene Symbol:
Dusp13
Gene Name:
dual specificity phosphatase 13
Gene Aliases:
-
Chromosome Location:
Chr.15: 2732897 - 2748457 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01537196_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dusp13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162408.1 NP_001155880.1 176 66
XM_008770545.1 XP_008768767.1 1 - 2 371 66
GenBank mRNA BC166532.1 - 176 7190084 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC166532.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500000128.rn.1 TC1500000017.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500000133.rn.1 TC1500000017.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500000129.rn.1 TC1500000017.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0