This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01537551_m1
See other SRD5A1 GE Assays ›
Gene Symbol:
Srd5a1
Gene Name:
steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1)
Gene Aliases:
S5AR 1
Chromosome Location:
Chr.1: 36320504 - 36354694 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01537551_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srd5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017070.3 NP_058766.2 441 67
GenBank mRNA BC127454.1 - 442 7313862 67
CO394005.1 - 441 67
J05035.1 - 294 67
S81448.1 - 314 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000376.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srd5a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

urogenital system development
liver development
steroid biosynthetic process
androgen biosynthetic process
androgen metabolic process
male gonad development
cellular response to starvation
response to organic cyclic compound
response to muscle activity
diterpenoid metabolic process
spinal cord development
hippocampus development
thalamus development
hypothalamus development
pituitary gland development
cerebral cortex development
cell differentiation
male genitalia development
female genitalia development
response to follicle-stimulating hormone
response to estradiol
cellular response to insulin stimulus
response to testosterone
serotonin metabolic process
progesterone metabolic process
response to drug
circadian sleep/wake cycle, REM sleep
response to estrogen
male sex differentiation
oxidation-reduction process
bone development
response to growth hormone
response to fungicide
cellular response to cAMP
cellular response to growth factor stimulus
cellular response to estradiol stimulus
cellular response to testosterone stimulus
cellular response to organic cyclic compound
cellular response to dexamethasone stimulus
cellular response to epinephrine stimulus
3-oxo-5-alpha-steroid 4-dehydrogenase activity
amide binding
cholestenone 5-alpha-reductase activity
NADPH binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100002246.rn.1 TC0100000376.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100002247.rn.1 TC0100000376.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0