This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01638043_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
Gene Aliases:
-
Chromosome Location:
Chr.18: 74156553 - 74164490 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01638043_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023093.1 NP_075581.1 11 - 12 1624 63
GenBank mRNA BC061830.1 - 11 - 12 1624 5599635 63
FQ214031.1 - 8 - 9 1036 63
FQ225416.1 - 6 - 7 939 63
FQ227531.1 - 6 - 7 939 63
FQ227720.1 - 5 - 6 759 63
J05266.1 - 11 - 12 1545 63
X56133.1 - 4 - 5 621 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
ATP metabolic process
hydrogen ion transmembrane transport
ATP binding
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P121101 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 79390 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 42845 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 538645 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 158626 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 658934 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 863769 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 137907 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121101 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P121101 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P121101 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P223817 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700371665 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 73881 - -
Agilent Rat cDNA Microarray (G4105A) 700371665 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10802861 - -
Affymetrix Rat Gene 1.0 ST Array 10802858 - -
Affymetrix Rat Gene 1.0 ST Array 10802857 - -
Affymetrix Rat Gene 1.0 ST Array 10802865 - -
Affymetrix Rat Gene 1.0 ST Array 10802854 - -
Affymetrix Rat Gene 1.0 ST Array 10802866 - -
Affymetrix Rat Gene 1.0 ST Array 10802855 - -
Affymetrix Rat Gene 1.0 ST Array 10802859 - -
Affymetrix Rat Gene 1.0 ST Array 10802862 - -
Affymetrix Rat Gene 1.0 ST Array 10802863 - -
Affymetrix Rat Gene 1.0 ST Array 10802860 - -
Affymetrix Rat Gene 1.0 ST Array 10802856 - -
Affymetrix Rat Gene 1.0 ST Array 10802864 - -
Affymetrix Rat Gene 1.1 ST Array 10802853 - -
Rat Genome 230 2.0 Array 1370378_at - -
Rat Genome 230 2.0 Array 1370378_PM_at - -
Affymetrix HT Rat Focus Array 1370378_at - -
Affymetrix HT Rat Focus Array 1370378_PM_at - -
Affymetrix Rat Clariom S Assay PSR1800006267.rn.1 TC1800000866.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800006266.rn.1 TC1800000866.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800006268.rn.1 TC1800000866.rn.1 RTA TC
Rat Expression Set 230 1370378_PM_at - -
Rat Expression Set 230 1370378_at - -
Affymetrix Rat Gene 1.0 ST Array 10802853 - -
Rat Genome U34 Set X56133_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21690537 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359244 - -
RatRef-12 Expression BeadChip ILMN_53344 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0