This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01638721_g1
See other IDI1 GE Assays ›
Gene Symbol:
Idi1
Gene Name:
isopentenyl-diphosphate delta isomerase 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 57976266 - 57984036 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01638721_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idi1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053539.2 NP_445991.2 4 - 5 769 117
GenBank mRNA AF003835.1 - 4 - 5 750 117
BC089786.1 - 4 - 5 769 7322086 117
FQ219635.1 - 4 - 5 418 117
FQ220448.1 - 4 - 5 416 117
FQ225062.1 - 4 - 5 416 117
FQ226929.1 - 4 - 5 422 117
FQ227874.1 - 4 - 5 419 117

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idi1

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
isoprenoid biosynthetic process
response to stilbenoid
dimethylallyl diphosphate biosynthetic process
magnesium ion binding
isopentenyl-diphosphate delta-isomerase activity
hydrolase activity
manganese ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P076638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 28453 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140707 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 652713 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 717965 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432839 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 165716 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076638 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076638 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076638 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P251944 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12843 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 59254 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10799247 - -
Affymetrix Rat Gene 1.0 ST Array 10799246 - -
Affymetrix Rat Gene 1.0 ST Array 10799244 - -
Affymetrix Rat Gene 1.0 ST Array 10799243 - -
Affymetrix Rat Gene 1.0 ST Array 10799242 - -
Affymetrix Rat Gene 1.0 ST Array 10799245 - -
Affymetrix Rat Gene 1.1 ST Array 10799241 - -
Rat Genome 230 2.0 Array 1388872_PM_at - -
Rat Genome 230 2.0 Array 1368878_at - -
Rat Genome 230 2.0 Array 1368878_PM_at - -
Rat Genome 230 2.0 Array 1388872_at - -
Affymetrix HT Rat Focus Array 1368878_PM_at - -
Affymetrix HT Rat Focus Array 1368878_at - -
Affymetrix Rat Clariom S Assay PSR1700013419.rn.1 TC1700001978.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700013422.rn.1 TC1700001978.rn.1 RTA TC
Rat Expression Set 230 1368878_PM_at - -
Rat Expression Set 230 1368878_at - -
Affymetrix Rat Gene 1.0 ST Array 10799241 - -
Rat Genome U34 Set rc_AI045395_at - -
Rat Genome U34 Set AF003835_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21364896 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1367519 - -
RatRef-12 Expression BeadChip ILMN_62987 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline