This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01646778_m1
See other ATOX1 GE Assays ›
Gene Symbol:
Atox1
Gene Name:
antioxidant 1 copper chaperone
Gene Aliases:
Atx1
Chromosome Location:
Chr.10: 40790850 - 40805886 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01646778_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053359.2 NP_445811.1 1 - 2 102 111
GenBank mRNA AF177671.1 - 1 - 2 117 111
BC058458.1 - 1 - 2 126 6919265 111
BI281905.1 - 3 - 4 111
FQ220104.1 - 1 - 2 103 111

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF177671.1, BC058458.1, NM_053359.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atox1

Gene Ontology Categories:

Function(s) Process(es)

copper ion transport
cellular copper ion homeostasis
response to oxidative stress
intracellular copper ion transport
copper ion transmembrane transport
copper ion transmembrane transporter activity
copper ion binding
copper chaperone activity
copper-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000041391.rn.1 TC1000004404.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000041393.rn.1 TC1000004404.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0