This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01648667_m1
See other TNFRSF11B GE Assays ›
Gene Symbol:
Tnfrsf11b
Gene Name:
tumor necrosis factor receptor superfamily, member 11b
Gene Aliases:
Opg
Chromosome Location:
Chr.7: 93798580 - 93826586 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01648667_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012870.2 NP_037002.1 2 - 3 508 100
GenBank mRNA BC081830.1 - 2 - 3 508 7096099 100
U94330.1 - 2 - 3 527 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf11b

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
signal transduction
response to nutrient
response to inorganic substance
extracellular matrix organization
response to magnesium ion
negative regulation of odontogenesis of dentin-containing tooth
response to drug
response to estrogen
negative regulation of bone resorption
response to arsenic-containing substance
molecular_function
cytokine activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0