This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01648668_m1
See other TNFRSF11B GE Assays ›
Gene Symbol:
Tnfrsf11b
Gene Name:
tumor necrosis factor receptor superfamily, member 11b
Gene Aliases:
Opg
Chromosome Location:
Chr.7: 93798580 - 93826586 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01648668_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012870.2 NP_037002.1 3 - 4 697 77
GenBank mRNA BC081830.1 - 3 - 4 697 7096099 77
U94330.1 - 3 - 4 716 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf11b

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
signal transduction
response to nutrient
response to inorganic substance
extracellular matrix organization
response to magnesium ion
negative regulation of odontogenesis of dentin-containing tooth
response to drug
response to estrogen
negative regulation of bone resorption
response to arsenic-containing substance
molecular_function
cytokine activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P458021 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 815595 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 250885 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 355130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 562080 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 458261 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 463610 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 429773 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 190754 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 952207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 635420 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 272423 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 634157 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 769790 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 108688 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 704718 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 75910 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 805863 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 912625 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 564421 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 154552 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 317930 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 143264 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 44423 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 27451 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P458021 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P458021 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P458021 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P458021 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 7445 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 73131 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10903730 - -
Affymetrix Rat Gene 1.0 ST Array 10903726 - -
Affymetrix Rat Gene 1.0 ST Array 10903727 - -
Affymetrix Rat Gene 1.0 ST Array 10903728 - -
Affymetrix Rat Gene 1.0 ST Array 10903729 - -
Affymetrix Rat Gene 1.1 ST Array 10903725 - -
Rat Genome 230 2.0 Array 1369407_PM_at - -
Rat Genome 230 2.0 Array 1369407_at - -
Affymetrix HT Rat Focus Array 1369407_PM_at - -
Affymetrix HT Rat Focus Array 1369407_at - -
Affymetrix Rat Clariom S Assay PSR0700024005.rn.1 TC0700003139.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700024003.rn.1 TC0700003139.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700024002.rn.1 TC0700003139.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700024004.rn.1 TC0700003139.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700024001.rn.1 TC0700003139.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700024000.rn.1 TC0700003139.rn.1 RTA TC
Rat Expression Set 230 1369407_PM_at - -
Rat Expression Set 230 1369407_at - -
Affymetrix Rat Gene 1.0 ST Array 10903725 - -
Rat Genome U34 Set U94330_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1374220 - -
RatRef-12 Expression BeadChip ILMN_60409 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0