This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01649127_m1
See other SLCO2B1 GE Assays ›
Gene Symbol:
Slco2b1
Gene Name:
solute carrier organic anion transporter family, member 2b1
Gene Aliases:
Slc21a9, moat1
Chromosome Location:
Chr.1: 164623575 - 164671619 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01649127_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco2b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080786.1 NP_542964.1 13 - 14 1996 73
XM_006229721.2 XP_006229783.1 13 - 14 2305 73
XM_006229722.2 XP_006229784.1 13 - 14 2302 73
XM_006229723.2 XP_006229785.1 13 - 14 3305 73
XM_006229724.2 XP_006229786.1 13 - 14 3276 73
XM_006229725.2 XP_006229787.1 13 - 14 3261 73
XM_006229727.2 XP_006229789.1 13 - 14 3273 73
XM_006229728.2 XP_006229790.1 13 - 14 3258 73
XM_008759668.1 XP_008757890.1 14 - 15 2095 73
GenBank mRNA AF169409.1 - 13 - 14 1996 73
AF169410.1 - 13 - 14 1956 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006121.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco2b1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

liver development
bile acid and bile salt transport
sodium-independent icosanoid transport
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
sodium-independent organic anion transmembrane transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P128007 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 564907 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 132453 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 741139 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 505753 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 122277 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 79076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 564258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 371560 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 48328 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 501109 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 592381 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 621977 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 337674 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 790368 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730022 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 349999 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 973763 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 301651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 148391 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 789879 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 251986 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 513530 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 424986 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 526821 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 471966 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 968945 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 859060 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 574343 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 300812 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 867975 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 51211 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 44200 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 108923 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 41237 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7900 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128007 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128007 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P128007 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491450 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128007 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 701881342 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700484415 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17698 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 95903 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 104450 - -
Agilent Rat cDNA Microarray (G4105A) 700484415 - -
Agilent Rat cDNA Microarray (G4105A) 701881342 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10723911 - -
Affymetrix Rat Gene 1.0 ST Array 10723912 - -
Affymetrix Rat Gene 1.0 ST Array 10723904 - -
Affymetrix Rat Gene 1.0 ST Array 10723915 - -
Affymetrix Rat Gene 1.0 ST Array 10723916 - -
Affymetrix Rat Gene 1.0 ST Array 10723905 - -
Affymetrix Rat Gene 1.0 ST Array 10723917 - -
Affymetrix Rat Gene 1.0 ST Array 10723906 - -
Affymetrix Rat Gene 1.0 ST Array 10723913 - -
Affymetrix Rat Gene 1.0 ST Array 10723907 - -
Affymetrix Rat Gene 1.0 ST Array 10723903 - -
Affymetrix Rat Gene 1.0 ST Array 10723910 - -
Affymetrix Rat Gene 1.0 ST Array 10723909 - -
Affymetrix Rat Gene 1.0 ST Array 10723908 - -
Affymetrix Rat Gene 1.0 ST Array 10723914 - -
Affymetrix Rat Gene 1.1 ST Array 10723902 - -
Rat Genome 230 2.0 Array 1368295_PM_at - -
Rat Genome 230 2.0 Array 1368296_at - -
Rat Genome 230 2.0 Array 1388288_PM_at - -
Rat Genome 230 2.0 Array 1388288_at - -
Rat Genome 230 2.0 Array 1368296_PM_at - -
Rat Genome 230 2.0 Array 1368295_at - -
Affymetrix HT Rat Focus Array 1368296_at - -
Affymetrix HT Rat Focus Array 1368296_PM_at - -
Affymetrix HT Rat Focus Array 1388288_PM_at - -
Affymetrix HT Rat Focus Array 1388288_at - -
Affymetrix HT Rat Focus Array 1368295_at - -
Affymetrix HT Rat Focus Array 1368295_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100048567.rn.1 TC0100006121.rn.1 RTA TC
Rat Expression Set 230 1388288_PM_at - -
Rat Expression Set 230 1368296_at - -
Rat Expression Set 230 1388288_at - -
Rat Expression Set 230 1368295_at - -
Rat Expression Set 230 1368295_PM_at - -
Rat Expression Set 230 1368296_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10723902 - -
Rat Genome U34 Set X61106cds_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22369487 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1358836 - -
RatRef-12 Expression BeadChip ILMN_64055 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline