This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01749516_g1
See other CPTP GE Assays ›
Gene Symbol:
Cptp
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
Gltpd1
Chromosome Location:
Chr.5: 173314228 - 173318384 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01749516_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cptp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007703.1 NP_001007704.1 2 - 3 403 91
XM_006239576.2 XP_006239638.1 2 - 3 564 91
GenBank mRNA BC083599.1 - 2 - 3 403 7133008 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500004215.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cptp

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P774468 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 114308 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 944355 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 864096 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 172099 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P774468 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P774468 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P774468 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P774468 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 90200 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10882237 - -
Affymetrix Rat Gene 1.0 ST Array 10882236 - -
Affymetrix Rat Gene 1.0 ST Array 10882235 - -
Affymetrix Rat Gene 1.1 ST Array 10882234 - -
Rat Genome 230 2.0 Array 1391437_PM_at - -
Rat Genome 230 2.0 Array 1391437_at - -
Affymetrix HT Rat Focus Array 1391437_at - -
Affymetrix HT Rat Focus Array 1391437_PM_at - -
Affymetrix Rat Clariom S Assay PSR0500032199.rn.1 TC0500004215.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500032201.rn.1 TC0500004215.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500032202.rn.1 TC0500004215.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500032205.rn.1 TC0500004215.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500032198.rn.1 TC0500004215.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500032200.rn.1 TC0500004215.rn.1 RTA TC
Rat Expression Set 230 1391437_PM_at - -
Rat Expression Set 230 1391437_at - -
Affymetrix Rat Gene 1.0 ST Array 10882234 - -
Rat Genome U34 Set rc_AA874995_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21546692 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1363911 - -
RatRef-12 Expression BeadChip ILMN_52985 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0