This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01750395_m1
See other PRSS22 GE Assays ›
Gene Symbol:
Prss22
Gene Name:
protease, serine, 22
Gene Aliases:
-
Chromosome Location:
Chr.10: 13370098 - 13374808 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01750395_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106984.1 NP_001100454.1 4 - 5 642 92
GenBank mRNA AJ005642.1 - 3 - 4 626 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000002375.rn.1

Set Membership:

Amplicon length between 86 and 100 Protease Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000020938.rn.1 TC1000002375.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0