This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01751022_g1
See other PRSS33 GE Assays ›
Gene Symbol:
Prss33
Gene Name:
protease, serine, 33
Gene Aliases:
-
Chromosome Location:
Chr.10: 13173509 - 13176288 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01751022_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109027.1 NP_001102497.1 5 - 6 694 71
XM_006245874.2 XP_006245936.1 6 - 7 984 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000002371.rn.1

Set Membership:

Amplicon length between 71 and 85 Protease 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
serine-type peptidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051054 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 26088 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432251 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051054 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051054 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051054 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10740926 - -
Affymetrix Rat Gene 1.0 ST Array 10740928 - -
Affymetrix Rat Gene 1.0 ST Array 10740925 - -
Affymetrix Rat Gene 1.0 ST Array 10740929 - -
Affymetrix Rat Gene 1.0 ST Array 10740927 - -
Affymetrix Rat Gene 1.1 ST Array 10740924 - -
Affymetrix Rat Clariom S Assay PSR1000020907.rn.1 TC1000002371.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000020905.rn.1 TC1000002371.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000020906.rn.1 TC1000002371.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10740924 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21780281 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1368591 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0