This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01752487_g1
See other H2AFV GE Assays ›
Gene Symbol:
H2afv
Gene Name:
H2A histone family, member V
Gene Aliases:
H2A.Z-2
Chromosome Location:
Chr.14: 86687782 - 86706626 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01752487_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H2afv
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106019.1 NP_001099489.1 1 - 2 89 124

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001106019.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Process(es)

DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1400016396.rn.1 TC1400002196.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400016400.rn.1 TC1400002196.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1400016402.rn.1 TC1400002196.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0