This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01753494_m1
See other MTRF1 GE Assays ›
Gene Symbol:
Mtrf1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 61661995 - 61689867 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01753494_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtrf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003751518.3 XP_003751566.1 8 - 9 1402 61
XM_006252367.2 XP_006252429.1 8 - 9 1440 61
XM_006252368.2 XP_006252430.1 7 - 8 1292 61
XM_006252369.2 XP_006252431.1 8 - 9 1402 61
XM_008770927.1 XP_008769149.1 8 - 9 1131 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

translation release factor

Gene Ontology Categories:

Function(s) Process(es)

translational termination
translation release factor activity, codon specific

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0