This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01756316_g1
See other ATP5B GE Assays ›
Gene Symbol:
Atp5b
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.7: 2504708 - 2511748 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01756316_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134364.1 NP_599191.1 9 - 10 1507 72
GenBank mRNA BC099743.1 - 9 - 10 1490 7937989 72
DQ403101.1 - 6 - 7 987 72
FQ214851.1 - 6 - 7 999 72
FQ216116.1 - 7 - 8 1089 72
FQ230301.1 - 7 - 8 1126 72
FQ231816.1 - 5 - 6 886 72
M19044.1 - 8 - 9 1323 72
M57634.1 - 6 - 7 972 72

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5b

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
osteoblast differentiation
lipid metabolic process
ATP biosynthetic process
receptor-mediated endocytosis
negative regulation of cell adhesion involved in substrate-bound cell migration
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
regulation of intracellular pH
calcium ion binding
ATP binding
ATPase activity
lipoprotein particle receptor activity
MHC class I protein binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P481516 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 801140 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 22195 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 632539 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 838665 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481516 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481516 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P481516 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P481516 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700492649 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700529925 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 26290 - -
Agilent Rat cDNA Microarray (G4105A) 700529925 - -
Agilent Rat cDNA Microarray (G4105A) 700492649 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10892974 - -
Affymetrix Rat Gene 1.0 ST Array 10892976 - -
Affymetrix Rat Gene 1.0 ST Array 10892971 - -
Affymetrix Rat Gene 1.0 ST Array 10892973 - -
Affymetrix Rat Gene 1.0 ST Array 10892969 - -
Affymetrix Rat Gene 1.0 ST Array 10892972 - -
Affymetrix Rat Gene 1.0 ST Array 10892975 - -
Affymetrix Rat Gene 1.0 ST Array 10892970 - -
Affymetrix Rat Gene 1.1 ST Array 10892968 - -
Rat Genome 230 2.0 Array 1370275_at - -
Rat Genome 230 2.0 Array 1370275_PM_at - -
Affymetrix HT Rat Focus Array 1370275_PM_at - -
Affymetrix HT Rat Focus Array 1370275_at - -
Affymetrix Rat Clariom S Assay PSR0700000164.rn.1 TC0700000018.rn.1 RTA TC
Rat Expression Set 230 1370275_PM_at - -
Rat Expression Set 230 1370275_at - -
Affymetrix Rat Gene 1.0 ST Array 10892968 - -
Rat Genome U34 Set rc_AI105050_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21464653 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1350324 - -
RatRef-12 Expression BeadChip ILMN_66749 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0