This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01756385_m1
See other GPR146 GE Assays ›
Gene Symbol:
Gpr146
Gene Name:
G protein-coupled receptor 146
Gene Aliases:
RGD1560731
Chromosome Location:
Chr.12: 17343396 - 17358628 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01756385_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109062.1 NP_001102532.1 1 - 2 105 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109062.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200010128.rn.1 TC1200001195.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200010133.rn.1 TC1200001195.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline