This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01756564_g1
See other AIMP2 GE Assays ›
Gene Symbol:
Aimp2
Gene Name:
aminoacyl tRNA synthetase complex-interacting multifunctional protein 2
Gene Aliases:
Jtv1
Chromosome Location:
Chr.12: 12738788 - 12748345 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01756564_g1
Size
Availability Inventoried
Catalog # 4331182
Price 272.00
Your Price
Online offer: 256.65
256.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aimp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037348.1 NP_001032425.1 3 - 4 612 70
XM_006248863.2 XP_006248925.1 3 - 4 559 70
GenBank mRNA BC107947.1 - 3 - 4 612 7386574 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aimp2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

translation
apoptotic process
negative regulation of cell proliferation
positive regulation of protein ubiquitination
Type II pneumocyte differentiation
positive regulation of neuron death

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P512460 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 668009 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 854981 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 340251 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 294025 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 963522 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 583033 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 638039 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 559126 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 111322 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 25237 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P512460 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P512460 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P512460 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P512460 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 21572 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10756696 - -
Affymetrix Rat Gene 1.0 ST Array 10756695 - -
Affymetrix Rat Gene 1.0 ST Array 10756694 - -
Affymetrix Rat Gene 1.0 ST Array 10756697 - -
Affymetrix Rat Gene 1.1 ST Array 10756692 - -
Rat Genome 230 2.0 Array 1371498_at - -
Rat Genome 230 2.0 Array 1371498_PM_at - -
Affymetrix HT Rat Focus Array 1371498_PM_at - -
Affymetrix HT Rat Focus Array 1371498_at - -
Affymetrix Rat Clariom S Assay PSR1200001530.rn.1 TC1200000217.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001534.rn.1 TC1200000217.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001533.rn.1 TC1200000217.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001532.rn.1 TC1200000217.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001535.rn.1 TC1200000217.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200001531.rn.1 TC1200000217.rn.1 RTA TC
Rat Expression Set 230 1371498_at - -
Rat Expression Set 230 1371498_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10756692 - -
Rat Genome U34 Set rc_AA799766_at - -
Rat Genome U34 Set rc_AA957244_f_at - -
Rat Genome U34 Set rc_AA957244_i_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21290704 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-szvnz:80/100.66.77.193:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0