This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01757171_m1
See other TNFRSF21 GE Assays ›
Gene Symbol:
Tnfrsf21
Gene Name:
tumor necrosis factor receptor superfamily, member 21
Gene Aliases:
-
Chromosome Location:
Chr.9: 20546159 - 20621051 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01757171_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108207.1 NP_001101677.1 3 - 4 1692 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001664.rn.1

Set Membership:

Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

B cell apoptotic process
adaptive immune response
apoptotic process
humoral immune response
negative regulation of B cell proliferation
negative regulation of myelination
negative regulation of T cell proliferation
myelination
regulation of oligodendrocyte differentiation
T cell receptor signaling pathway
neuron apoptotic process
cellular response to tumor necrosis factor
oligodendrocyte apoptotic process
negative regulation of interleukin-5 secretion
negative regulation of interleukin-13 secretion
negative regulation of interleukin-10 secretion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900013696.rn.1 TC0900001664.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0