This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01757173_m1
See other TNFRSF21 GE Assays ›
Gene Symbol:
Tnfrsf21
Gene Name:
tumor necrosis factor receptor superfamily, member 21
Gene Aliases:
-
Chromosome Location:
Chr.9: 20546159 - 20621051 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01757173_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108207.1 NP_001101677.1 5 - 6 2188 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900001664.rn.1

Set Membership:

Amplicon length between 86 and 100 Apoptosis 3' Most Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

B cell apoptotic process
adaptive immune response
apoptotic process
humoral immune response
negative regulation of B cell proliferation
negative regulation of myelination
negative regulation of T cell proliferation
myelination
regulation of oligodendrocyte differentiation
T cell receptor signaling pathway
neuron apoptotic process
cellular response to tumor necrosis factor
oligodendrocyte apoptotic process
negative regulation of interleukin-5 secretion
negative regulation of interleukin-13 secretion
negative regulation of interleukin-10 secretion

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P185441 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P459212 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 673231 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826265 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 239090 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723257 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 794498 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 586310 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 768030 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 89540 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 619017 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 247296 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 510459 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 844434 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 389403 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 66906 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 3298 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 515034 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 617516 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 209816 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 802029 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84979 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 619458 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 901194 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 454153 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 707901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224666 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 618611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 949208 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 678483 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 516035 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 97703 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 207939 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202763 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 514426 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 602226 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 282471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 681494 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 111522 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 677847 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894379 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323997 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 555398 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 570914 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 424689 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 360354 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 464176 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 584700 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 317924 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 960206 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469198 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 623000 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 39906 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 566258 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 254874 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 145884 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 512760 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 788854 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 69205 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 14785 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 167828 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 27066 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 64392 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 75626 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 108033 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 60783 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112734 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P185441 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P459212 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P459212 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P185441 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P459212 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P185441 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P185441 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 70710 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 55499 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10926742 - -
Affymetrix Rat Gene 1.0 ST Array 10926744 - -
Affymetrix Rat Gene 1.0 ST Array 10926743 - -
Affymetrix Rat Gene 1.0 ST Array 10926741 - -
Affymetrix Rat Gene 1.0 ST Array 10926745 - -
Affymetrix Rat Gene 1.0 ST Array 10926746 - -
Affymetrix Rat Gene 1.1 ST Array 10926740 - -
Rat Genome 230 2.0 Array 1393584_PM_at - -
Rat Genome 230 2.0 Array 1393584_at - -
Rat Genome 230 2.0 Array 1391573_PM_at - -
Rat Genome 230 2.0 Array 1391573_at - -
Affymetrix HT Rat Focus Array 1391573_at - -
Affymetrix HT Rat Focus Array 1391573_PM_at - -
Affymetrix HT Rat Focus Array 1393584_at - -
Affymetrix HT Rat Focus Array 1393584_PM_at - -
Affymetrix Rat Clariom S Assay PSR0900013694.rn.1 TC0900001664.rn.1 RTA TC
Rat Expression Set 230 1391573_PM_at - -
Rat Expression Set 230 1391573_at - -
Rat Expression Set 230 1393584_PM_at - -
Rat Expression Set 230 1393584_at - -
Affymetrix Rat Gene 1.0 ST Array 10926740 - -
Rat Genome U34 Set rc_AA891842_g_at - -
Rat Genome U34 Set rc_AA891842_at - -
Rat Genome U34 Set rc_AI045201_at - -
Rat Genome U34 Set rc_AI172520_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21245871 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1370428 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0