This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01758494_m1
See other GFI1B GE Assays ›
Gene Symbol:
Gfi1b
Gene Name:
growth factor independent 1B transcription repressor
Gene Aliases:
-
Chromosome Location:
Chr.3: 7190721 - 7203444 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01758494_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gfi1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107823.1 NP_001101293.1 1 - 2 89 81
XM_006233789.1 XP_006233851.1 1 - 2 128 81
XM_008761646.1 XP_008759868.1 1 - 2 128 81
XM_008761651.1 XP_008759873.1 1 - 2 128 81
GenBank mRNA BC158685.1 - 1 - 2 104 8374780 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC158685.1, NM_001107823.1, XM_006233789.1, XM_008761646.1, XM_008761651.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
hemopoiesis
positive regulation of granulocyte differentiation
regulation of erythrocyte differentiation
regulation of histone H3-K4 methylation
negative regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II transcription factor binding
nucleic acid binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300021194.rn.1 TC0300002786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300021189.rn.1 TC0300002786.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-hnx4s:80/100.66.72.123:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0