This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01759399_m1
See other RNMT GE Assays ›
Gene Symbol:
Rnmt
Gene Name:
RNA (guanine-7-) methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.18: 64084773 - 64108246 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01759399_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008299.2 NP_001008300.1 10 - 11 1440 76
XM_006254894.1 XP_006254956.1 10 - 11 1601 76
XM_006254895.2 XP_006254957.1 10 - 11 2875 76
XM_006254896.2 XP_006254958.1 9 - 10 1297 76
GenBank mRNA BC085858.1 - 10 - 11 1440 7190303 76
CN543861.1 - 1 - 2 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800000753.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnmt

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
biological_process
RNA (guanine-N7)-methylation
molecular_function
RNA binding
mRNA (guanine-N7-)-methyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P849067 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1015355 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 427655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740816 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 208782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 870845 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 616962 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 904044 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 353426 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 362904 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 170701 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 483254 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 405235 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 368557 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 540947 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153671 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 546038 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 54181 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 89460 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015355 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P849067 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015355 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P849067 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P849067 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1015355 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P849067 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 76543 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 58986 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 79712 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 59495 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 90917 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 96884 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10805065 - -
Affymetrix Rat Gene 1.0 ST Array 10805059 - -
Affymetrix Rat Gene 1.0 ST Array 10805063 - -
Affymetrix Rat Gene 1.0 ST Array 10805053 - -
Affymetrix Rat Gene 1.0 ST Array 10805058 - -
Affymetrix Rat Gene 1.0 ST Array 10805056 - -
Affymetrix Rat Gene 1.0 ST Array 10805055 - -
Affymetrix Rat Gene 1.0 ST Array 10805060 - -
Affymetrix Rat Gene 1.0 ST Array 10805057 - -
Affymetrix Rat Gene 1.0 ST Array 10805061 - -
Affymetrix Rat Gene 1.0 ST Array 10805062 - -
Affymetrix Rat Gene 1.0 ST Array 10805054 - -
Affymetrix Rat Gene 1.0 ST Array 10805064 - -
Affymetrix Rat Gene 1.1 ST Array 10805052 - -
Rat Genome 230 2.0 Array 1372867_at - -
Rat Genome 230 2.0 Array 1398572_PM_at - -
Rat Genome 230 2.0 Array 1372867_PM_at - -
Rat Genome 230 2.0 Array 1398572_at - -
Affymetrix HT Rat Focus Array 1398572_PM_at - -
Affymetrix HT Rat Focus Array 1398572_at - -
Affymetrix HT Rat Focus Array 1372867_at - -
Affymetrix HT Rat Focus Array 1372867_PM_at - -
Affymetrix Rat Clariom S Assay PSR1800005445.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005438.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005446.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005448.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005439.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005440.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005447.rn.1 TC1800000753.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800005436.rn.1 TC1800000753.rn.1 RTA TC
Rat Expression Set 230 1372867_PM_at - -
Rat Expression Set 230 1398572_PM_at - -
Rat Expression Set 230 1398572_at - -
Rat Expression Set 230 1372867_at - -
Affymetrix Rat Gene 1.0 ST Array 10805052 - -
Rat Genome U34 Set rc_AA800016_at - -
Rat Genome U34 Set rc_AI177333_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20856282 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1365236 - -
RatRef-12 Expression BeadChip ILMN_64230 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0