This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01761015_g1
See other NDUFA2 GE Assays ›
Gene Symbol:
Ndufa2
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 29585671 - 29587760 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01761015_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106153.1 NP_001099623.1 1 - 2 121 99
GenBank mRNA FQ212326.1 - 1 - 2 97 99
FQ216914.1 - 1 - 2 99 99
FQ216957.1 - 1 - 2 116 99
FQ217727.1 - 1 - 2 95 99
FQ223951.1 - 1 - 2 96 99
FQ224026.1 - 1 - 2 99 99

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1800009140.rn.1 TC1800001274.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800009141.rn.1 TC1800001274.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0