This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01762516_m1
See other DDX19A GE Assays ›
Gene Symbol:
Ddx19a Ddx19b
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
DEAD (Asp-Glu-Ala-As) box polypeptide 19B
Gene Aliases:
Ddx19, ZD10B
Zd10a
Chromosome Location:
Chr.19: 43251510 - 43271866 on Build Rnor_6.0
Chromosome Location:
Chr.19: 43217199 - 43246196 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01762516_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx19a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY303540.1 - 129 83
Gene Symbol: Ddx19b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005895.2 NP_001005895.2 1 - 2 196 83
GenBank mRNA AY751756.1 - 1 - 2 129 83
EV774940.1 - 1 - 2 196 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000604.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx19a
Click here to find siRNAs for Gene Ddx19b

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
nucleic acid binding
helicase activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1900005138.rn.1 TC1900000604.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900005137.rn.1 TC1900000604.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900005136.rn.1 TC1900000604.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline