This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01764169_g1
See other SIGLEC10 GE Assays ›
Gene Symbol:
Siglec10
Gene Name:
sialic acid binding Ig-like lectin 10
Gene Aliases:
Siglecg
Chromosome Location:
Chr.1: 98512663 - 98521740 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01764169_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Siglec10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106250.1 NP_001099720.1 2 - 3 205 81
XM_006228948.2 XP_006229010.1 2 - 3 239 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001106250.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100042845.rn.1 TC0100005309.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline