This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01765029_g1
See other NUDT5 GE Assays ›
Gene Symbol:
Nudt5
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 5
Gene Aliases:
-
Chromosome Location:
Chr.17: 76387027 - 76410313 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01765029_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007733.1 NP_001007734.1 3 - 4 278 108
XM_008771875.1 XP_008770097.1 3 - 4 297 108
GenBank mRNA BC079176.1 - 3 - 4 278 7130276 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt5

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
ribonucleoside diphosphate catabolic process
magnesium ion binding
nucleoside-diphosphatase activity
ADP-sugar diphosphatase activity
snoRNA binding
ADP-ribose diphosphatase activity
m7G(5')pppN diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700014696.rn.1 TC1700002199.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700014697.rn.1 TC1700002199.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0