This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01765164_m1
See other ECHDC3 GE Assays ›
Gene Symbol:
Echdc3
Gene Name:
enoyl CoA hydratase domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 76002304 - 76022813 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01765164_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101010.1 NP_001094480.1 3 - 4 636 113
GenBank mRNA BC101897.1 - 3 - 4 636 7932594 113
FQ219501.1 - 3 - 4 674 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700000981.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P169327 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P169327 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P169327 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P169327 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P169333 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P169327 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1373921_PM_at - -
Rat Genome 230 2.0 Array 1373921_at - -
Affymetrix HT Rat Focus Array 1373921_at - -
Affymetrix HT Rat Focus Array 1373921_PM_at - -
Affymetrix Rat Clariom S Assay PSR1700006705.rn.1 TC1700000981.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700006710.rn.1 TC1700000981.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700006707.rn.1 TC1700000981.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700006711.rn.1 TC1700000981.rn.1 RTA TC
Rat Expression Set 230 1373921_PM_at - -
Rat Expression Set 230 1373921_at - -
Rat Genome U34 Set rc_AI169007_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0