This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01765865_m1
See other TMPRSS11E GE Assays ›
Gene Symbol:
Tmprss11e
Gene Name:
transmembrane protease, serine 11e
Gene Aliases:
RGD1561698
Chromosome Location:
Chr.14: 23069213 - 23113258 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01765865_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008770102.1 XP_008768324.1 10 - 11 1225 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Protease 3' Most Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cognition
serine-type endopeptidase activity
serine-type peptidase activity

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026993 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026993 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026993 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026993 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P295104 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21624899 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371647 - -
RatRef-12 Expression BeadChip ILMN_64746 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0