This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01766566_g1
See other ISLR GE Assays ›
Gene Symbol:
Islr
Gene Name:
immunoglobulin superfamily containing leucine-rich repeat
Gene Aliases:
-
Chromosome Location:
Chr.8: 62946732 - 62949680 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01766566_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Islr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001126300.1 NP_001119772.1 1 - 2 93 77
GenBank mRNA BC160841.1 - 1 - 2 93 7461347 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC160841.1, NM_001126300.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10944 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10944 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10944 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10944 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10944 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1389020_at - -
Rat Genome 230 2.0 Array 1389020_PM_at - -
Affymetrix HT Rat Focus Array 1389020_at - -
Affymetrix HT Rat Focus Array 1389020_PM_at - -
Affymetrix Rat Clariom S Assay PSR0800021500.rn.1 TC0800002646.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800021502.rn.1 TC0800002646.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800021499.rn.1 TC0800002646.rn.1 RTA TC
Rat Expression Set 230 1389020_at - -
Rat Expression Set 230 1389020_PM_at - -
Rat Genome U34 Set rc_AI172159_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21830139 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline