This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01767910_m1
See other TSFM GE Assays ›
Gene Symbol:
Tsfm
Gene Name:
Ts translation elongation factor, mitochondrial
Gene Aliases:
2A3-2, EF-Ts, EF-TsMt
Chromosome Location:
Chr.7: 70311948 - 70319346 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01767910_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tsfm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276476.1 NP_001263405.1 3 - 4 370 96
GenBank mRNA AJ005161.1 - 3 - 4 371 96
FQ220263.1 - 3 - 4 370 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700002902.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
translation elongation factor activity
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072442 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072442 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072442 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072442 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402435 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 702235629 - -
Agilent Rat cDNA Microarray (G4105A) 702235629 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1398603_PM_at - -
Rat Genome 230 2.0 Array 1398603_at - -
Affymetrix Rat Clariom S Assay PSR0700022269.rn.1 TC0700002902.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700022272.rn.1 TC0700002902.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700022273.rn.1 TC0700002902.rn.1 RTA TC
Rat Expression Set 230 1398603_at - -
Rat Expression Set 230 1398603_PM_at - -
Rat Genome U34 Set rc_AI104501_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21914764 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0