This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01771871_m1
See other DBX1 GE Assays ›
Gene Symbol:
Dbx1
Gene Name:
developing brain homeobox 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 104969218 - 104973648 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01771871_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dbx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009644.1 NP_001009644.1 1 - 2 370 92
GenBank mRNA AB197138.1 - 1 - 2 370 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100005475.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dbx1

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
ventral spinal cord interneuron specification
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100044823.rn.1 TC0100005475.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100044824.rn.1 TC0100005475.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline