This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01772746_m1
See other CES1A GE Assays ›
Gene Symbol:
Ces1a
Gene Name:
carboxylesterase 1A
Gene Aliases:
-
Chromosome Location:
Chr.19: 15339152 - 15366079 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01772746_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ces1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190375.1 NP_001177304.1 12 - 13 1448 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1900000156.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

esterase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
carboxylic ester hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068930 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141469 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P141469 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068930 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P141469 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068930 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1900001328.rn.1 TC1900000156.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900001327.rn.1 TC1900000156.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900001329.rn.1 TC1900000156.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21534546 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline