This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02087462_s1
See other CMTR2 GE Assays ›
Gene Symbol:
Cmtr2
Gene Name:
cap methyltransferase 2
Gene Aliases:
Ftsjd1, RGD1309394
Chromosome Location:
Chr.19: 41426536 - 41433367 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02087462_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cmtr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106186.1 NP_001099656.1 2 3167 104

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
cap2 mRNA methylation
mRNA (nucleoside-2'-O-)-methyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011609 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 941739 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 781184 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 173445 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 306828 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 219428 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 632209 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 71369 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011610 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P581181 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011610 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P581181 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P581181 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011610 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P581181 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P124027 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10807842 - -
Affymetrix Rat Gene 1.1 ST Array 10807841 - -
Affymetrix Rat Clariom S Assay PSR1900011571.rn.1 TC1900001417.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900011572.rn.1 TC1900001417.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1900011570.rn.1 TC1900001417.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10807841 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21800714 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1355002 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0