This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02106240_s1
See other GLTP GE Assays ›
Gene Symbol:
Gltp
Gene Name:
glycolipid transfer protein
Gene Aliases:
RGD1564442
Chromosome Location:
Chr.12: 47667556 - 47687708 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02106240_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gltp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134413.1 NP_001127885.1 5 1233 140
GenBank mRNA BC158884.1 - 5 1233 7938728 140

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycolipid transport
lipid binding
glycolipid transporter activity
glycolipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P592205 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 866647 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 699915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 318124 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793256 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 509982 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 491418 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 540819 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 726007 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773724 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 472761 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 887782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 715728 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 250515 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 677662 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 20763 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 14941 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P592205 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P592205 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P592205 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P505199 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P592205 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 37989 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10759095 - -
Affymetrix Rat Gene 1.0 ST Array 10759091 - -
Affymetrix Rat Gene 1.0 ST Array 10759094 - -
Affymetrix Rat Gene 1.0 ST Array 10759093 - -
Affymetrix Rat Gene 1.0 ST Array 10759092 - -
Affymetrix Rat Gene 1.1 ST Array 10759089 - -
Rat Genome 230 2.0 Array 1388480_at - -
Rat Genome 230 2.0 Array 1388480_PM_at - -
Affymetrix HT Rat Focus Array 1388480_PM_at - -
Affymetrix HT Rat Focus Array 1388480_at - -
Affymetrix Rat Clariom S Assay PSR1200007038.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007039.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007044.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007037.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007043.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007041.rn.1 TC1200000805.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200007042.rn.1 TC1200000805.rn.1 RTA TC
Rat Expression Set 230 1388480_PM_at - -
Rat Expression Set 230 1388480_at - -
Affymetrix Rat Gene 1.0 ST Array 10759089 - -
Rat Genome U34 Set rc_AI008713_at - -
Rat Genome U34 Set rc_AA900632_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22129344 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1369554 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline