This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02345863_g1
See other BCS1L GE Assays ›
Gene Symbol:
Bcs1l
Gene Name:
BC1 (ubiquinol-cytochrome c reductase) synthesis-like
Gene Aliases:
-
Chromosome Location:
Chr.9: 81868265 - 81872198 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02345863_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcs1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007666.1 NP_001007667.1 2 - 3 625 140
GenBank mRNA BC083660.1 - 2 - 3 625 7135331 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900000904.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcs1l

Gene Ontology Categories:

Function(s) Process(es)

mitochondrion organization
biological_process
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
mitochondrial respiratory chain complex III assembly
molecular_function
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900007496.rn.1 TC0900000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900007495.rn.1 TC0900000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900007492.rn.1 TC0900000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900007497.rn.1 TC0900000904.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0