This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02347257_m1
See other GLYAT GE Assays ›
Gene Symbol:
Glyat
Gene Name:
glycine-N-acyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.1: 229060129 - 229081113 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02347257_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glyat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009648.2 NP_001009648.1 3 - 4 315 94
XM_006231121.2 XP_006231183.1 5 - 6 622 94
XM_006231122.2 XP_006231184.1 4 - 5 537 94
XM_006231123.2 XP_006231185.1 4 - 5 359 94
XM_008760257.1 XP_008758479.1 5 - 6 1502 94
GenBank mRNA AB201944.1 - 3 - 4 315 94
BC087043.1 - 4 - 5 413 7108337 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003253.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glyat

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycine metabolic process
response to toxic substance
monocarboxylic acid metabolic process
glycine N-acyltransferase activity
glycine N-benzoyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100026883.rn.1 TC0100003253.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100026884.rn.1 TC0100003253.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0