This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02395362_m1
See other DHRS7 GE Assays ›
Gene Symbol:
Dhrs7
Gene Name:
dehydrogenase/reductase (SDR family) member 7
Gene Aliases:
DR-NR#3
Chromosome Location:
Chr.6: 95480042 - 95502845 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02395362_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013098.1 NP_001013116.1 1 - 2 240 66
GenBank mRNA AB108671.1 - 1 - 2 240 66
BC104713.1 - 1 - 2 146 7370181 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600002645.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhrs7

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267628 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P267628 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P267628 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267628 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600018844.rn.1 TC0600002645.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600018845.rn.1 TC0600002645.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600018843.rn.1 TC0600002645.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0