This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02395972_m1
See other GPR139 GE Assays ›
Gene Symbol:
Gpr139
Gene Name:
G protein-coupled receptor 139
Gene Aliases:
-
Chromosome Location:
Chr.1: 188854371 - 188895223 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02395972_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr139
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024241.1 NP_001019412.1 1 - 2 99 70
GenBank mRNA AB196531.1 - 1 - 2 99 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006503.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr139

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
neuropeptide receptor activity
protein dimerization activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098818 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 341196 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 606807 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853028 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 16313 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 378999 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 133714 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 843263 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 436711 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 691505 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 956580 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 938450 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 147131 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573763 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 333076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325772 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 363286 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 123547 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 422708 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 571903 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 254337 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 617613 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 603316 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 302407 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 898550 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 847093 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 925377 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 214494 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 307346 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163563 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 904920 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 951470 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 290770 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 873759 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 825739 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 142577 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 136497 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 22799 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 54764 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 111391 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098818 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098818 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098818 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503745 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 27551 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 43085 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 12073 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10725287 - -
Affymetrix Rat Gene 1.0 ST Array 10725288 - -
Affymetrix Rat Gene 1.1 ST Array 10725286 - -
Affymetrix Rat Clariom S Assay PSR0100051059.rn.1 TC0100006503.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100051060.rn.1 TC0100006503.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10725286 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1530481 - -
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360837 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0