This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn02758935_s1
See other GPR146 GE Assays ›
Gene Symbol:
Gpr146
Gene Name:
G protein-coupled receptor 146
Gene Aliases:
RGD1560731
Chromosome Location:
Chr.12: 17343396 - 17358628 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn02758935_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109062.1 NP_001102532.1 2 960 63
XM_006248955.2 XP_006249017.1 2 3105 63
XM_006248956.2 XP_006249018.1 2 1439 63
XM_006248957.2 XP_006249019.1 2 973 63
XM_006248958.2 XP_006249020.1 3 2713 63

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P852017 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852017 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P852017 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P852017 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P852017 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10760664 - -
Affymetrix Rat Gene 1.1 ST Array 10760663 - -
Rat Genome 230 2.0 Array 1373158_at - -
Rat Genome 230 2.0 Array 1373158_PM_at - -
Rat Genome 230 2.0 Array 1382266_PM_at - -
Rat Genome 230 2.0 Array 1382266_at - -
Affymetrix HT Rat Focus Array 1373158_PM_at - -
Affymetrix HT Rat Focus Array 1382266_at - -
Affymetrix HT Rat Focus Array 1373158_at - -
Affymetrix HT Rat Focus Array 1382266_PM_at - -
Rat Expression Set 230 1373158_at - -
Rat Expression Set 230 1373158_PM_at - -
Rat Expression Set 230 1382266_at - -
Rat Expression Set 230 1382266_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10760663 - -
Rat Genome U34 Set rc_AI170446_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1368702 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline