This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn03020912_m1
See other PPP1R14C GE Assays ›
Gene Symbol:
Ppp1r14c
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 14c
Gene Aliases:
Kepi
Chromosome Location:
Chr.1: 39811314 - 39923071 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn03020912_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r14c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133425.3 NP_596916.1 1 - 2 904 103
GenBank mRNA AF407168.3 - 1 - 2 904 103
BC086978.1 - 1 - 2 377 7191557 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000397.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r14c

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphorylation
negative regulation of catalytic activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100002385.rn.1 TC0100000397.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100002387.rn.1 TC0100000397.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100002386.rn.1 TC0100000397.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100002384.rn.1 TC0100000397.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0